Learn More
Hopkins Medical Products NEUROPEN NEUROTIPS
SDP
Catalog No. NC0232457
Encompass_Preferred
Single-Use Neurotips •Sterile •Single Use •Semi-Sharp To Reduce The Risk Of Skin Punctures •Ensures A Quantifiable Force Of 40G Can Be Exerted Safely Onto Skin •Red & White Neurotips Provide A Contrast When Used For Peripheral Vision Testing •100/Bx
Catalog No. NC0232457 Supplier Hopkins Medical Products Supplier No. 595405
Single-Use Neurotips •Sterile •Single Use •Semi-Sharp To Reduce The Risk Of Skin Punctures •Ensures A Quantifiable Force Of 40G Can Be Exerted Safely Onto Skin •Red & White Neurotips Provide A Contrast When Used For Peripheral Vision Testing •100/Bx