Learn More
CPC Scientific H-Glu-Ile-Gly-Asp-Glu-Glu-Asn-Ser-Ala-Lys-Phe-Pro-Ile-Gly-Arg-Arg-Asp-Phe-Asp-Met-Leu-Arg-Cys-Met-Leu-Gly-Arg-Val-Tyr-Arg-Pro-Cys-Trp-Gln-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MCH003A
Encompass
Neuropeptide EI-Gly-Arg-Arg-MCH (human, mouse, rat) has been shown to be more potent in stimulating feeding in the rat than MCH following intracerebroventricular administration although it did not exhibit better agonist activity in in vitro assays. Its enhanced activity in feeding behaviour compared to MCH might be due to its reduced susceptibility to proteolytic degradation by MCH-degrading enzymes such as endopeptidase 24.11 and aminopeptidase M.ONE-LETTER SEQUENCE: EIGDEENSAKFPIGRRDFDMLRCMLGRVYRPCWQVMOLECULAR FORMULA: C182H282N54O52S4MOLECULAR WEIGHT:4186.84STORAGE CONDITIONS: -20 5CRESEARCH AREA: HormonalREFERENCES: A.Viale et al., J. Biol. Chem., 274, 6536 (1999); L.Maulon-Feraille et al., J. Pharmacol. Exp. Ther., 302, 766 (2002)
Catalog No. 50-210-9723