Learn More
Medchemexpress LLC Neuropeptide FF (NPFF) | 99566-27-5 | 99.4% | 1081.27 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP12481MG
Encompass_Preferred
Neuropeptide FF (NPFF) is an octapeptide from the RF-amide family, acting as an agonist for NPFF1 and NPFF2 receptors. It is involved in diverse physiological processes, including abstinence syndrome, memory, and pain modulation. Present in the mammalian central nervous system and periphery, NPFF is key for research into opioid tolerance, analgesia, and various pain conditions.
  • Agonist for NPFF1 and NPFF2 receptors
  • Induces abstinence syndrome
  • Exerts antiopioid and analgesic effects
  • Regulates memory, autonomic, neuroendocrine functions
  • Modulates pain and opioid antinociception
  • Reduces food and stimulates water intake
  • Alters cardiovascular parameters
  • Research tool for opioid tolerance and morphine-induced analgesia
  • Applied in studies of hypertension, nociception, and pain types
Catalog No. 50-004-24802