Learn More
CPC Scientific H-Ser-Phe-Arg-Asn-Gly-Val-Gly-Thr-Gly-Met-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific NEUP010B
Encompass
This peptide almost completely mimics the effects of the full length peptide (Neuropeptide S (human). ONE-LETTER SEQUENCE: SFRNGVGTGMMOLECULAR FORMULA: C42H68N14O14S1MOLECULAR WEIGHT:1025.15STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [904910-39-0], RESEARCH AREA: NeuropeptidesREFERENCES: A.L.Roth et al., J. Biol. Chem., 281, 20809 (2006)
Catalog No. 50-210-9899