Learn More
Eurofins Discoverx Neuropeptide W-23, 1 mg. (92-1175)
Supplier:  Eurofins Discoverx 921175
Encompass
Neuropeptide W-23 (NPW-23) is a 23 amino acid long neuropeptide corresponding to the N terminus of Neuropeptide W. It is a mediator of the neuroendocrine system with functions in feeding control. Found in the hypothalamus, hippocampus. Agonist of the neuropeptide B and W receptors (NPBW1 and NPBW2). Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9703