Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neuropeptide y (13-36), amide, human | 122341-40-6 | 98.8% | 3000.40 | 10 MG
SDP

Catalog No. 5000424876
Encompass_Preferred

Neuropeptide Y (13-36), amide, human is a selective neuropeptide Y2 receptor agonist. This product is for research use only and is not sold to patients.

  • Selective neuropeptide Y2 receptor agonist
  • Purity of 98.8%
  • Molecular weight of 3000.40
  • Appearance: white to off-white solid
  • Impairs social novelty preference in naive mice (in vivo)
  • Increases mean arterial blood pressure in male rats (in vivo)

Catalog No. 50-004-24876 Supplier Medchemexpress LLC Supplier No. HYP148010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neuropeptide Y (13-36), amide, human is a selective neuropeptide Y2 receptor agonist. This product is for research use only and is not sold to patients.

  • Selective neuropeptide Y2 receptor agonist
  • Purity of 98.8%
  • Molecular weight of 3000.40
  • Appearance: white to off-white solid
  • Impairs social novelty preference in naive mice (in vivo)
  • Increases mean arterial blood pressure in male rats (in vivo)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.