Learn More
Medchemexpress LLC Neuropeptide Y (13-36), porcine | 113662-54-7 | 99.79% | 2982.36 | 1 MG
SDP
Catalog No. 5000423680
Encompass_Preferred
Neuropeptide Y (13-36), porcine is a selective neuropeptide Y2 receptor agonist.
  • Selective neuropeptide Y2 receptor agonist
Catalog No. 50-004-23680 Supplier Medchemexpress LLC Supplier No. HYP10111MG
Neuropeptide Y (13-36), porcine is a selective neuropeptide Y2 receptor agonist.
  • Selective neuropeptide Y2 receptor agonist