Learn More
Medchemexpress LLC Neuropeptide Y (18-36) (porcine) | 114495-97-5 | 98.8% | 2456.80 | 10 MG
SDP
Catalog No. 5000397386
Encompass_Preferred
Neuropeptide Y (18-36) (porcine) is a competitive neuropeptide Y (NPY) cardiac receptor antagonist. It inhibits the binding of I-NPY to cardiac ventricular membranes in a concentration-dependent manner with an IC50 value of 158 nM and a Ki value of 140 nM. It can be used for the research of congestive heart failure.
  • Competitive neuropeptide Y (NPY) cardiac receptor antagonist
  • Inhibits I-NPY binding with IC50 of 158 nM and Ki of 140 nM
  • Shifts inhibitory adenylate cyclase activity dose-response curve of NPY to the right
  • Abolishes effect of NPY on adenylate cyclase activity
  • Used for research of congestive heart failure
Catalog No. 50-003-97386 Supplier Medchemexpress LLC Supplier No. HYP367810MG
Neuropeptide Y (18-36) (porcine) is a competitive neuropeptide Y (NPY) cardiac receptor antagonist. It inhibits the binding of I-NPY to cardiac ventricular membranes in a concentration-dependent manner with an IC50 value of 158 nM and a Ki value of 140 nM. It can be used for the research of congestive heart failure.
  • Competitive neuropeptide Y (NPY) cardiac receptor antagonist
  • Inhibits I-NPY binding with IC50 of 158 nM and Ki of 140 nM
  • Shifts inhibitory adenylate cyclase activity dose-response curve of NPY to the right
  • Abolishes effect of NPY on adenylate cyclase activity
  • Used for research of congestive heart failure