Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neuropeptide Y (18-36) (porcine) | 114495-97-5 | 98.8% | 2456.80 | 5 MG
SDP

Catalog No. 5000399483
Encompass_Preferred

Neuropeptide Y (18-36) (porcine) is a competitive neuropeptide Y (NPY) cardiac receptor antagonist. It inhibits the binding of I-NPY to cardiac ventricular membranes in a concentration-dependent manner. With an IC50 of 158 nM and a Ki of 140 nM for I-NPY, it is suitable for research on congestive heart failure.

  • Competitive NPY cardiac receptor antagonist
  • Inhibits I-NPY binding to cardiac ventricular membranes
  • IC50: 158 nM (I-NPY), Ki: 140 nM (I-NPY)
  • Can be used for research of congestive heart failure
  • Appearance: solid
  • Color: white to off-white
  • Molecular formula: C112H174N36O27

Catalog No. 50-003-99483 Supplier Medchemexpress LLC Supplier No. HYP36785MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neuropeptide Y (18-36) (porcine) is a competitive neuropeptide Y (NPY) cardiac receptor antagonist. It inhibits the binding of I-NPY to cardiac ventricular membranes in a concentration-dependent manner. With an IC50 of 158 nM and a Ki of 140 nM for I-NPY, it is suitable for research on congestive heart failure.

  • Competitive NPY cardiac receptor antagonist
  • Inhibits I-NPY binding to cardiac ventricular membranes
  • IC50: 158 nM (I-NPY), Ki: 140 nM (I-NPY)
  • Can be used for research of congestive heart failure
  • Appearance: solid
  • Color: white to off-white
  • Molecular formula: C112H174N36O27

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.