Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neuropeptide Y(29-64) | 303052-45-1 | 99.9% | 4272.70 | 10 MG
SDP

Catalog No. 5000424490
Encompass_Preferred

Neuropeptide Y(29-64) is a 36 amino acid peptide that serves as a fragment of Neuropeptide Y. It is intended for research use only.

  • Target: Neuropeptide Y receptor
  • Pathway: GPCR/G protein, neuronal signaling
  • Soluble in water (≥ 46.67 mg/mL) and DMSO (16.67 mg/mL)
  • Appearance: Solid, white to off-white

Catalog No. 50-004-24490 Supplier Medchemexpress LLC Supplier No. HYP160110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neuropeptide Y(29-64) is a 36 amino acid peptide that serves as a fragment of Neuropeptide Y. It is intended for research use only.

  • Target: Neuropeptide Y receptor
  • Pathway: GPCR/G protein, neuronal signaling
  • Soluble in water (≥ 46.67 mg/mL) and DMSO (16.67 mg/mL)
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.