Learn More
Bachem Neuropeptide Y (3-36) (human, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4031095.1000
Encompass_Preferred
Neuropeptide Y (3-36) (human, rat) Trifluoroacetate, H-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2, 150138-78-6, C175H269N53O54S, Mr 4011.45, 1mg. Main degradation product of NPY in serum and Y2/Y5 receptor agonist.
Catalog No. 50-259-740