Learn More
CPC Scientific H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP104A
Encompass
NPY fragment that has a slight effect on NPY-induced inhibition of the vas deferens response to electrical stimulation. ONE-LETTER SEQUENCE: YPSKPDNPGEDAPAEDMARYYSAL-NH2MOLECULAR FORMULA: C116H170N30O40S1MOLECULAR WEIGHT:2656.87STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [131448-51-6], SYNONYMS: Neuropeptide Y (1-24) amide (human, rat)RESEARCH AREA: NeuropeptidesREFERENCES: J.C. Martel et al., Mol. Pharmacol., 38, 494 (1990)
Catalog No. 50-210-9931