Learn More
Bachem Neuropeptide Y (human, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4012616.0500
Encompass_Preferred
Neuropeptide Y (human, rat) Trifluoroacetate, H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2, 90880-35-6, C189H285N55O57S, Mr 4271.74, 0.5mg. NPY (human, rat).
Catalog No. NC1138585