Learn More
CPC Scientific H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP106A
Encompass
Brain peptide that possesses both gastrointestinal and vasoconstrictor properties.ONE-LETTER SEQUENCE: YPSKPDNPGEDAPAEDLARYYSALRHYINLITRQRY-NH2MOLECULAR FORMULA: C190H287N55O57MOLECULAR WEIGHT:4253.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [83589-17-7], SYNONYMS: NPY (porcine), Neuropeptide Y (porcine)RESEARCH AREA: NeuropeptidesREFERENCES: K. Tatemoto et al., PNAS, 79, 5485 (1982)
Catalog No. 50-210-9935