Learn More
Medchemexpress LLC HY-P1601 1mg Medchemexpress, Neuropeptide Y(29-64) CAS:303052-45-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP16011MG
Encompass_Preferred
Medchemexpress, HY-P1601 1mg Neuropeptide Y(29-64) CAS:303052-45-1 Neuropeptide Y(29-64) is a 36 amino acid peptide, a fragment of Neuropeptide Y. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3071