Learn More
ACROBiosystems ACROBiosystems Human Neuropilin-1 / NRP1 / CD304 Protein, Fc Tag
Catalog No. 501957228
Encompass_Preferred
Recombinant Protein;1MG;Human NRP1, Fc Tag (NR1-H5252) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96.5 kDa. The protein migrates as 105-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Human NRP1, Fc Tag (Cat. No. NR1-H5252) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-7228 Supplier ACROBiosystems Supplier No. NR1H52521MG
Recombinant Protein;1MG;Human NRP1, Fc Tag (NR1-H5252) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96.5 kDa. The protein migrates as 105-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Human NRP1, Fc Tag (Cat. No. NR1-H5252) with a linear range of 0.2-3 ng/mL (QC tested).