Learn More
ACROBiosystems ACROBiosystems Mouse Neuropilin-1 / NRP1 / CD304 Protein, Mouse IgG2a Fc Tag, low endotoxin
Catalog No. 501957235
Encompass_Preferred
Recombinant Protein;1MG;Mouse NRP1, Mouse IgG2a Fc Tag, low endotoxin (NR1-M5254) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Pro 856 (Accession # P97333-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 120.5 kDa. The protein migrates as 125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse NRP1, Mouse IgG2a Fc Tag (Cat. No. NR1-M5254) with a linear range of 0.5-4 ng/mL (QC tested).
Catalog No. 50-195-7235 Supplier ACROBiosystems Supplier No. NR1M52541MG
Recombinant Protein;1MG;Mouse NRP1, Mouse IgG2a Fc Tag, low endotoxin (NR1-M5254) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Pro 856 (Accession # P97333-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 120.5 kDa. The protein migrates as 125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse NRP1, Mouse IgG2a Fc Tag (Cat. No. NR1-M5254) with a linear range of 0.5-4 ng/mL (QC tested).