Learn More
Sino Biological Human Neuropilin-1 / NRP1 / CD304 Protein (His & AVI Tag), Biotinylated 20ug
Supplier:  Sino Biological 10011H27HB20
Encompass_Preferred
A DNA sequence encoding the human NRP1 (NP_001019799.1) (Met1-Lys644) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5203