Learn More
ACROBiosystems Recombinant Protein;Human Neuropilin-1 / NRP1 / CD304 Protein;HEK293;1MG;NR1,NRP1,Neuropilin-1,NRP,VEGF165R,CD304,NRP-1
Catalog No. 501658456
Encompass_Preferred
Recombinant Protein;Human Neuropilin-1 / NRP1 / CD304 Protein;1MG;Human NRP1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.7 kDa. The protein migrates as 85-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NRP1, His Tag (Cat. No. NR1-H5228) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF165, Tag Free (Cat. No. VE5-H8210) with a linear range of 1-8 ng/mL (QC tested).
Catalog No. 50-165-8456 Supplier ACROBiosystems Supplier No. NR1H52281MG
Add to Cart
Add to Cart
Recombinant Protein;Human Neuropilin-1 / NRP1 / CD304 Protein;1MG;Human NRP1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.7 kDa. The protein migrates as 85-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NRP1, His Tag (Cat. No. NR1-H5228) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF165, Tag Free (Cat. No. VE5-H8210) with a linear range of 1-8 ng/mL (QC tested).