Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Neuropilin-1 / NRP1 / CD304 Protein, His Tag & Avi Tag (Avitag™);HEK293;200UG;NR1,NRP1,Neuropilin-1,NRP,VEGF165R,CD304,NRP-1
Supplier:  ACROBiosystems NR1H82E3200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human Neuropilin-1 / NRP1 / CD304 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human NRP1, His Tag (NR1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # O14786-1(V179A)).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 73.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human NRP1, His Tag (Cat. No. NR1-H82E3) with a linear range of 0.5-16 ng/mL (QC tested).
Catalog No. 50-165-8152