Learn More
Sigma Aldrich Fine Chemicals Biosciences NeuroPorterTM Transfection
Catalog No. 502908083
Encompass
Neuroporter(TM) Transfection Reagent is a unique formulation of a proprietary cationic lipid optimized for delivery of DNA into primary neurons glial cells and cultured neuronal cell lines with high efficiency and low toxicity. The Neuroporter(TM) Transfection Kit was designed for difficult-to-transfect primary neurons addressing past problems such as poor cell viability low transfection efficiency and neuro-degeneration.
Catalog No. 50-290-8083 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NPT011KT
Neuroporter(TM) Transfection Reagent is a unique formulation of a proprietary cationic lipid optimized for delivery of DNA into primary neurons glial cells and cultured neuronal cell lines with high efficiency and low toxicity. The Neuroporter(TM) Transfection Kit was designed for difficult-to-transfect primary neurons addressing past problems such as poor cell viability low transfection efficiency and neuro-degeneration.