Learn More
Hamilton™ Neuros™ 700/1700 Series Adapter Kit
Catalog No. 14785714 Shop All Hamilton Company Products
Change view
Click to view available options
Needle Gauge:
33 G
Catalog No. Needle Gauge
14-785-714 33 G
1 options
Catalog No. 14-785-714 Supplier Hamilton™ Supplier No. 6546004
Add to Cart
Add to Cart
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Needle Gauge 33 G
Includes Adapter, two sleeves, and a six pack of 33 gauge needles
Product Type Adapter Kit
For Use With (Equipment) Removable Needle Syringe (100μL and Smaller)