Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ NEUROS™ Syringes
SDP

Catalog No. 10000923 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
10 μL, 33 gauge
100 μL, 33 gauge
2 μL, 30 gauge
25 μL, 33 gauge
5 μL, 33 gauge
5 to 100 μL, 33 gauge
50 μL, 33 gauge
Includes:
6/pk.
Adapter Kit
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
10-000-923 5 μL, 33 gauge Adapter Kit
10-000-888 100 μL, 33 gauge Adapter Kit
10-000-895 5 to 100 μL, 33 gauge 6/pk.
10-000-922 2 μL, 30 gauge
10-000-924 10 μL, 33 gauge Adapter Kit
10-000-925 25 μL, 33 gauge Adapter Kit
10-000-926 50 μL, 33 gauge Adapter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-000-923 Supplier Stoelting™ Supplier No. 53496
Add to Cart
Add to Cart

Stoelting™ Hamilton Neuros™ syringe technology provides functionality for controlled animal injections.

  • Accurately dispenses 50 nL to 100 μL of liquid through an ultrafine needle protected by a custom-designed sleeve
  • Developed specifically for the neurosciences
  • Minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location
  • Flexible options for a variety of uses
  • Crafted to have very little to no dead volume which preserves the maximum amount of buffer and solution.
  • Blunt point (point style 3) ensures the sample is delivered directly to the site
  • Two different needle sleeves
  • Sleeve with a blind stop is ideal for cannulated applications and ensures targeted administration with an adjustable penetration depth
  • Version without a blind stop works best with stereotaxic holders
  • Both types provide an adjustable needle exposure of 0 to 20 mm

Specifications

For Use With (Application) Neuroscience
Size 5 μL, 33 gauge
Model Blunt
Includes Adapter Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.