Learn More
Medchemexpress LLC HY-P7270 10ug Medchemexpress, Neuroserpin (CHO-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP727010UG
Encompass_Preferred
Medchemexpress, HY-P7270 10ug Neuroserpin (CHO-expressed), Human CAS: Recombinant Human Neuroserpin (CHO-expressed) is a member of the serpin family of serine protease inhibitors, playing an important role in synaptic plasticity and memory formation in the hippocampus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4372