Learn More
Medchemexpress LLC HY-P0234 1mg , Neurotensin CAS:39379-15-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02341MG
Encompass_Preferred
HY-P0234 1mg Neurotensin CAS: 39379-15-2 Neurotensin, a gut tridecapeptide, acts as a potent cellular mitogen for various colorectal and pancreatic cancers which possess high-affinity neurotensin receptors (NTR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1565