Learn More
Bachem Neurotensin Acetate
SDP
Supplier:  Bachem 4006469.0025
Encompass_Preferred
Neurotensin Acetate, Pyr-Leu-Tyr-Glu-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu-OH, 55508-42-4, C78H121N21O20, Mr 1672.95, 25mg. The tridecapeptide hormone neurotensin (NT) has a wide spectrum of pharmacological effects.
Catalog No. 50-259-846