Learn More
CPC Scientific H-Arg-Arg-Pro-Tyr-Ile-Leu-OH 1MG
SDP
Supplier:  CPC Scientific NEUP181A
Encompass
SEQUENCE: H-Arg-Arg-Pro-Tyr-Ile-Leu-OHONE-LETTER SEQUENCE: RRPYILMOLECULAR FORMULA: C38H64N12O8MOLECULAR WEIGHT: 817.00STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [60482-95-3]SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:S. St. Pierre et al., J. Med. Chem., 24, 370 (1981)SKU(s): NEUP-181A, NEUP-181B, NEUP-181C
Catalog No. 50-210-9959