Learn More
Medchemexpress LLC HY-P0251 1mg Medchemexpress, Neurotensin(8-13) CAS:60482-95-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02511MG
Encompass_Preferred
Medchemexpress, HY-P0251 1mg Neurotensin(8-13) CAS:60482-95-3 Neurotensin (8-13) is an active fragment of Neurotensin,. Neurotensin(8-13) results in a decrease in cell-surface NT1 receptors (NTR1) density. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4079