Learn More
Medchemexpress LLC Neurotoxin Inhibitor | 951571-70-3 | 99.0% | 346.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11272210MM/1ML
Encompass_Preferred
Neurotoxin Inhibitor is a neurotoxin inhibitor that promotes the expression of DJ-1 protein, reduces the level of oxidative stress, and thereby protects dopaminergic neurons. It can be used for the study of Parkinson's disease.
  • Promotes the expression of DJ-1 protein.
  • Reduces the level of oxidative stress.
  • Protects dopaminergic neurons.
  • Can be used for the study of Parkinson's disease.
  • Dose-dependently upregulates the expression of DJ-1 protein in SH-SY5Y cells and primary cortical neurons of mice.
  • Effectively alleviates excessive oxidative stress in SH-SY5Y cells treated with MPP⁺.
Catalog No. 50-004-37913