Learn More
Creative Biomart Botulinum Toxin Type B
Catalog No. 502084079
Encompass_Preferred
Neurotoxin produced by fermentation of clostridium botulinum type B. The protein exists in noncovalent association with hemagglutinin and nonhemagglutinin proteins as a neurotoxin complex. The neurotoxin complex is recovered from the fermentation process and purified through a series of precipitation and chromatography steps.
Catalog No. 50-208-4079 Supplier Creative Biomart Supplier No. THP0028
Neurotoxin produced by fermentation of clostridium botulinum type B. The protein exists in noncovalent association with hemagglutinin and nonhemagglutinin proteins as a neurotoxin complex. The neurotoxin complex is recovered from the fermentation process and purified through a series of precipitation and chromatography steps.