Learn More
Molecular Devices Molecular Devices Neurotransmitter Uptake Assay Kit, Fluorescence, Live-Cell, 96/384, 50-Plate
Catalog No. 5000014584
Encompass_Preferred
The Molecular Devices Neurotransmitter Transporter Uptake Assay Kit is a homogeneous, fluorescence-based live-cell assay for measuring uptake activity of serotonin (SERT), norepinephrine (NET), and dopamine (DAT) transporters. The kit uses a fluorescent substrate and masking dye to enable no-wash, mix-and-read endpoint or kinetic measurements on bottom-read microplate readers.
  • Homogeneous, no-wash live-cell assay format.
  • Fluorescence-based detection suitable for kinetic or endpoint reads.
  • Compatible with 96-well and 384-well black clear-bottom plates.
  • Bulk kit contains reagents for 50 plates (5 vials of lyophilized dye/masking mix).
  • Reconstituted dye solution stable for limited time; store frozen reagents at -20°C.
Catalog No. 50-000-14584 Supplier Molecular Devices Supplier No. R8174
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices Neurotransmitter Transporter Uptake Assay Kit is a homogeneous, fluorescence-based live-cell assay for measuring uptake activity of serotonin (SERT), norepinephrine (NET), and dopamine (DAT) transporters. The kit uses a fluorescent substrate and masking dye to enable no-wash, mix-and-read endpoint or kinetic measurements on bottom-read microplate readers.
  • Homogeneous, no-wash live-cell assay format.
  • Fluorescence-based detection suitable for kinetic or endpoint reads.
  • Compatible with 96-well and 384-well black clear-bottom plates.
  • Bulk kit contains reagents for 50 plates (5 vials of lyophilized dye/masking mix).
  • Reconstituted dye solution stable for limited time; store frozen reagents at -20°C.