Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices IMAP FP Phosphodiesterase Kit, 384-well, 800 data points

Catalog No. 5000014467
Encompass_Preferred

IMAP FP Phosphodiesterase Evaluation Kit provides reagents for measuring phosphodiesterase activity using fluorescence polarization detection. The kit is supplied in an evaluation format and contains IMAP binding reagent, binding buffers, and labeled cyclic nucleotide substrates sufficient for multiple assays in a 384-well plate format.

  • Intended use: phosphodiesterase fluorescence polarization assays.
  • Contains IMAP binding reagent and binding buffers.
  • Includes labeled cAMP and cGMP substrates.
  • Throughput: approximately 800 data points in a standard 384-well plate.
  • Detection method: fluorescence polarization compatible reagents.

Catalog No. 50-000-14467 Supplier Molecular Devices Supplier No. R8175
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IMAP FP Phosphodiesterase Evaluation Kit provides reagents for measuring phosphodiesterase activity using fluorescence polarization detection. The kit is supplied in an evaluation format and contains IMAP binding reagent, binding buffers, and labeled cyclic nucleotide substrates sufficient for multiple assays in a 384-well plate format.

  • Intended use: phosphodiesterase fluorescence polarization assays.
  • Contains IMAP binding reagent and binding buffers.
  • Includes labeled cAMP and cGMP substrates.
  • Throughput: approximately 800 data points in a standard 384-well plate.
  • Detection method: fluorescence polarization compatible reagents.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.