Learn More
Molecular Devices Molecular Devices Calcium Assay Kit, Fura-2 QBT, Homogeneous, No-Wash
Catalog No. 5000014602
Encompass_Preferred
The Fura-2 QBT Calcium sample kit is a homogeneous, mix-and-read assay designed to measure intracellular calcium mobilization using the ratiometric dye Fura-2. Proprietary quench/masking chemistry enables a no-wash protocol to reduce steps and minimize assay artifacts.
  • Homogeneous mix-and-read assay for calcium mobilization.
  • No-wash protocol to reduce assay steps and variability.
  • Ratiometric Fura-2 dye provides excitation at 340/380 nm, emission at 510 nm.
  • Sample kit format contains two plates worth of reagent.
  • Compatible with plate-based fluorescence readers and FLIPR systems.
  • Minimizes effects of uneven dye loading and dye leakage on results.
Catalog No. 50-000-14602 Supplier Molecular Devices Supplier No. R6139
Add to Cart
Add to Cart
The Fura-2 QBT Calcium sample kit is a homogeneous, mix-and-read assay designed to measure intracellular calcium mobilization using the ratiometric dye Fura-2. Proprietary quench/masking chemistry enables a no-wash protocol to reduce steps and minimize assay artifacts.
  • Homogeneous mix-and-read assay for calcium mobilization.
  • No-wash protocol to reduce assay steps and variability.
  • Ratiometric Fura-2 dye provides excitation at 340/380 nm, emission at 510 nm.
  • Sample kit format contains two plates worth of reagent.
  • Compatible with plate-based fluorescence readers and FLIPR systems.
  • Minimizes effects of uneven dye loading and dye leakage on results.