Learn More
Enzo Life Sciences SCREEN-WELL Neurotransmitter library (10 plate set) (100µl/well)
SDP
Supplier:  Enzo Life Sciences BML28100100
Encompass_Preferred
The SCREEN-WELL neurotransmitter library contains 661 CNS receptor ligands in a 96-well format. Ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, validating new assays, and for routine pharmacological applications. Includes 13 classes of receptor ligands in 10 deep-well plates: AdrenergicsHistaminergics(Melatonin Ligands) DopaminergicsIonotropic Glutamatergics SerotonergicsMetabotropic Glutamatergics Opioids(Sigma ligands)GABAergics CholinergicsPurinergics (Adenosines) Each ligand is supplied in solution in a biocompatible solvent at a standard concentration of 10mM (Opioid peptides are supplied at a concentrations of 1mM). Plates are available as a complete set, or individually. Supporting data are included. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0186
Add to Cart
Request a Quote