Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Neutralization Number Solvent for TAN (ASTM D3242, D664, D974)(4L)
SDP

Catalog No. NC9685563
Encompass_Preferred

Neutralization Number Solvent for TAN (ASTM D3242, D664, D974)(4L)

Catalog No. NC9685563 Supplier Aqua Solutions Supplier No. 65204L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neutralization Number Solvent for TAN (ASTM D3242, D664, D974)(4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.