Learn More
Neutec Group BioTool Swiss Dosi-Pump DP1000 Agar and Liquid Pump
SDP
Catalog No. 14910322 Shop All Neutec Group Products
Change view
Click to view available options
Dosi Pump DP1000
Catalog No.
14-910-322 Dosi Pump DP1000
1 options
Catalog No. 14-910-322 Supplier Neutec Group Supplier No. BT730000
Only null left
Add to Cart
Add to Cart
Request a Quote
State-of-the-art technology is suitable for various applications for liquid dispensing and agar filling
  • Easy to use software operates with a touch screen
  • Anti-Drop, drip suppression by retraction of pump movement
  • Dual head pump for smoother liquid / agar filling
  • Pump offers high flow rates of up to 1,000 ml/min.
  • Easy volume and speed calibration
  • Backlight graphics display for easy reading
Specifications
Product Type Dosi Filler Pump
Weight (English) 15.65 in.
Weight (Metric) 7.1 kg