Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutec Group Smart Dilutor
GSA_VASDP

Catalog No. 14910316 Shop All Neutec Group Products
Change view
Click to view available options
Model:
Single Pump
Two Pump
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
14-910-316 Two Pump
14-910-315 Single Pump
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-910-316 Supplier Neutec Group Supplier No. 90007901
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

Both systems include polyethylene bag-holder, two sets of tubing, filters, 5L bottles, power cable, and an instruction manual. 14-910-315 is also supplied with two pumps.

For dilution of solid, semi-solid, or paste types of microbiological samples directly into sterile homogenizer bags

Since these types of samples require a gravimetric type of dilution process, the Smart Dilutor has an integrated balance at the heart of the system. The operator simply places an unknown amount of sample in a sterile bag and the unit takes an initial weight of the sample, then the operator moves the lever and the microprocessor dispenses an amount of diluent by weight, to achieve the programmed dilution ratio. The system constantly monitors the weight in the sample bag to achieve a highly accurate dilution result. Once the programmed dilution ratio has been reached, the system notifies the operator that the process has finished.

  • Software also allows the instrument to be used as a dispensing device for a previously determined quantity of a liquid into any kind of container
  • Bar code provides traceability of the dilution process
  • Can be expanded to dilute up to six different buffers with the use of an optional accessory
  • Adjustable, multilevel arm enables dilution of various type containers
  • Tubing set and the bottles are fully autoclavable
  • Changing operation of tubing set is performed in less than 15 seconds

Certifications

Each device is calibrated and certified against equipment whose calibration is traceable to NIST™

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Pipet Dispensing System
Autoclavable Autoclavable
Dilution 0.1 g
Volume (Metric) 1 μL to 25 mL
For Use With (Application) Dilution of solid, semi solid, or paste types of microbiological samples directly into sterile homogenizer bags
Includes Polyethylene bag holder, two sets of tubing, filters, 5L bottles, power cable, instruction manual, two pump
Model Two Pump
Speed 950 mL/min.
Temperature (English) 41°F to 104°F
Temperature (Metric) 5°C to 40°C
Dimensions (L x W x H) 7 x 6.13 x 13.13 in. (178 x 156 x 333 mm)
Length (English) 7 in.
Length (Metric) 178 mm
Width (English) 6.13 in.
Width (Metric) 156 mm
Height (English) 13.13 in.
Height (Metric) 333 mm
Weight (English) 5.2 lb.
Weight (Metric) 6500 g
Electrical Requirements 85/264VAC 50/60Hz
Voltage 85/264 VAC
Wattage 40 w
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.