Learn More
Electron Microscopy Sciences Neutra-Pads™ Aldehyde Control
SDP
Catalog No. 5030013
Encompass
This product offers a safe, efficient and economical solution for the control of aldehyde wastes and/or spills, eliminating the exposure to potentially hazardous conditions. Neutra is a revolutionary grossing pad that absorbs and neutralizes 10% formalin, significantly reducing the effects of noxious vapors. The unique pad design encapsulates formalin and promotes neutralization. The exclusive plastic pad backing prevents escape of harmful liquids, keeping grossing area drier, cleaner and safer.
Each pad measures 20.3 x 25.4 cm (8" x 10").
Catalog No. 50-300-13 Supplier Electron Microscopy Sciences Supplier No. 6333201
This product offers a safe, efficient and economical solution for the control of aldehyde wastes and/or spills, eliminating the exposure to potentially hazardous conditions. Neutra is a revolutionary grossing pad that absorbs and neutralizes 10% formalin, significantly reducing the effects of noxious vapors. The unique pad design encapsulates formalin and promotes neutralization. The exclusive plastic pad backing prevents escape of harmful liquids, keeping grossing area drier, cleaner and safer.
Each pad measures 20.3 x 25.4 cm (8" x 10").