Learn More
Andwin Scientific NEUTRA-WIPES 4-ROLL PACK EA
SDP
Catalog No. 50727473
Encompass_Preferred
Useful for absorbing and neutralizing small formalin spills, Neutra-Wipes also double as liner for tissue trays and storage shelves.This includes 4 rolls or 240 wipes.
This item is not returnable. View return policy
Catalog No. 50-727-473 Supplier Andwin Scientific Supplier No. 6333404
Useful for absorbing and neutralizing small formalin spills, Neutra-Wipes also double as liner for tissue trays and storage shelves.This includes 4 rolls or 240 wipes.