Learn More
Creative Diagnostics NeutraAb Human Anti-HPIV3 Monoclonal Antibody, IgG, 200 μg, 1 mg, Unconjugated, Liquid, Neutralization, ELISA
Supplier:  Creative Diagnostics CABTNS1270
Encompass
NeutraAb™ Human Anti-HPIV3 Monoclonal Antibody is an unconjugated IgG antibody specifically designed for the detection and neutralization of Human Parainfluenza Virus type 3 (HPIV3). This antibody serves as a crucial reagent in infectious disease research, proving useful in various applications including neutralization assays and ELISA, offering high specificity and consistent results.
  • Unconjugated IgG antibody
  • Developed for HPIV3 detection and neutralization
  • Suitable for neutralization assays and ELISA
  • High specificity and consistent results
  • Available in liquid format
  • Product sizes: 200 μg, 1 mg
  • Alternative names include HPIV 3 and Human parainfluenza virus type 3
Catalog No. NC2406505