Learn More
Creative Diagnostics NeutraAb Mouse Anti-HPIV3 Monoclonal Antibody, 200 μg, clone PIA174, unconjugated, IgG, for ELISA and neutralization assays
Supplier:  Creative Diagnostics CABTNS1268
Encompass
This NeutraAb Mouse Anti-HPIV3 Monoclonal Antibody is a high-quality, unconjugated reagent designed for the detection and neutralization of Human Parainfluenza Virus Type 3 (HPIV3). The antibody is suitable for various applications including neutralization assays and ELISA, making it essential for research and diagnostic purposes related to HPIV3.
  • High-quality monoclonal antibody
  • Produced in mouse
  • Targets human parainfluenza virus type 3 (HPIV3)
  • Suitable for neutralization assays and ELISA
  • Unconjugated format
  • Available in 200 μg and 1 mg sizes
Catalog No. NC2406504