Learn More
StatLab™ NeutraGreen Aldehyde Test Strips
SDP
Catalog No. STLNTS50
Change view
Click to view available options
Packaging Quantity:
50 Strips/Pk.
Catalog No. Packaging Quantity
STLNTS50 50 Strips/Pk.
1 options
Catalog No. STLNTS50 Supplier StatLab™ Supplier No. NTS50
Add to Cart
Add to Cart
A quick dip providing color coded results is an effective and simple way to test with these aldehyde test strips.
  • Used to help determine if waste formalin has been safely and effectively neutralized for drain disposal
Specifications
Material Paper
Type Formalin Test Kit
Packaging Quantity 50 Strips/Pk.