Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ NeutraGreen™ Liquid Neutralizer
GSA_VASDP

Catalog No. STL15251G
Change view
Click to view available options
Format:
Bottle
Cube
Packaging Quantity:
1 gal.
2.5 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Packaging Quantity
STL15251G Cube 1 gal.
STL152518 Bottle 5 gal.
STL152525 Cube 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STL15251G Supplier StatLab™ Supplier No. 15251G
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.

Transforms waste formalin into an odorless, 100% non-hazardous material.

  • Comes in liquid form with no powders to mix or dissolve
  • Waste reaction time of 2 to 3 hr enables same day disposal
  • Third-party environmental data confirms NeutraGreen™ will neutralize waste formalin to a material that is non-toxic and non-hazardous
  • Test with NeutraGreen™ Test Strips to ensure waste has been properly neutralized for drain disposal

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity (English) 1 gal.
Capacity (Metric) 3.79 L
Material Pre-treated Formalin Neutralizer
Type Formalin Neutralizer
Format Cube
For Use With (Application) Neutralizing Waste Formalin
Packaging Quantity 1 gal.
Weight (English) 7.5 lb.
Weight (Metric) 3.4 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.