Learn More
Kem Medical Products Corp Kem-Safe™ Glutaraldehyde/OPA Neutralizing Solution, 12-32oz bottles/case
SDP
Catalog No. NC0377551
Encompass_Preferred
Eliminate toxic vapors created during disposal of waste solution. Will not stain soaking containers, sinks, etc. Prevents black staining. Neutralization is complete within 5 minutes and solution is ready for safe disposal. Safe for use in automatic scope washers. Each bottle will neutralize two (2) gallons of waste Glutaraldehyde/OPA solution.
This item is not returnable. View return policy
Catalog No. NC0377551 Supplier Kem Medical Products Corp Supplier No. 9074
Eliminate toxic vapors created during disposal of waste solution. Will not stain soaking containers, sinks, etc. Prevents black staining. Neutralization is complete within 5 minutes and solution is ready for safe disposal. Safe for use in automatic scope washers. Each bottle will neutralize two (2) gallons of waste Glutaraldehyde/OPA solution.