Learn More
Proteintech Group Inc NeutraKine IL-3 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc 690041IG100UG
Encompass_Preferred
The NeutraKine IL-3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine IL-3. This antibody recognizes human antigen. The NeutraKine IL-3 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Catalog No. 50-225-6529