Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VION™ Neutral Buffered Formalin 10% Prefilled Containers
SCI_ED

Catalog No. S001205 Shop All Aldon Products
Change view
Click to view available options
Quantity:
20 mL
60 mL
90 mL
120 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001205 20 mL
S001206 60 mL
S001204 120 mL
S001203 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S001205 Supplier VION™ Supplier No. VNFF026620C
Add to Cart
Add to Cart

Ensure specimen integrity from the bench to the lab with VION pre filled neutral buffered formalin containers, industry standard for secure tissue fixation.

  • Leak-Proof Security: Engineered to eliminate the risks of manual filling and hazardous leaks, these high-clarity polypropylene containers feature a specialized, dual-thread Tite-Rite™ seal specifically designed to protect both your staff and your most critical samples during pneumatic tube transport and rigorous shipping.
  • Gold-Standard Preservation: Pre-filled with VION's Histology Grade 10% Neutral Buffered Formalin to ensure consistent pH and superior morphological detail for immunohistochemistry and molecular testing.
  • Safety and Compliance: Shatter-resistant, pre-labeled containers reduce chemical exposure and meet all GHS labeling requirements for immediate use.
  • Specifications:
    • Container
    • Container Material: Polypropylene
    • Cap Material: Polyethylene
    • Contents Appearance: Clear, Colorless Liquid
    • pH: 7.0 ± 0.1
  • Note: This product can expose you to chemicals including Methanol and Formaldehyde, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Line VION Histology
Product Type Tissue Preservative
Quantity 20 mL
Solution Type Formalin
pH 7
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.