Learn More
Description
- Leak-Proof Security: Engineered to eliminate the risks of manual filling and hazardous leaks, these high-clarity polypropylene containers feature a specialized, dual-thread Tite-Rite™ seal specifically designed to protect both your staff and your most critical samples during pneumatic tube transport and rigorous shipping.
- Gold-Standard Preservation: Pre-filled with VION's Histology Grade 10% Neutral Buffered Formalin to ensure consistent pH and superior morphological detail for immunohistochemistry and molecular testing.
- Safety and Compliance: Shatter-resistant, pre-labeled containers reduce chemical exposure and meet all GHS labeling requirements for immediate use.
- Specifications:
- Container
- Container Material: Polypropylene
- Cap Material: Polyethylene
- Contents Appearance: Clear, Colorless Liquid
- pH: 7.0 ± 0.1
- Note: This product can expose you to chemicals including Methanol and Formaldehyde, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Histological |
| Product Line | VION Histology |
| Product Type | Tissue Preservative |
| Quantity | 60 mL |
| Solution Type | Formalin |
| pH | 7 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
