Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Neutral Cleaner Concentrate

Catalog No. 17623905 Shop All 3M Products
Change view
Click to view available options
Volume (English):
2.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-623-905 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-905 Supplier 3M™ Supplier No. 7010342466
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

No rinse, low foaming neutral cleaner concentrate won't harm washable hard floors or leave soapy residue that can dull or soften floor finish. Use with mop or automatic scrubber. Fresh scent. Dilution rate: 1:391. For use with Scotchgard™ Hard Floor Protection Products.

  • Low foaming in high-flow applications
  • Can be used with high-flow dispensing systems to fill auto-scrubbers
  • Light yellow with green tint
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleaner Concentrate
Form Liquid
Appearance Yellow
For Use With (Equipment) Mop or Automatic Scrubber
Quantity 2/Cs.
Volume (English) 2.5 gal.
Volume (Metric) 9.464 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.