Learn More
MSC Dow Corning 4098499 3 oz Cartridge Clear RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737
Supplier:  MSC 00231902
Encompass
Dow Corning 4098499 3 oz Cartridge Clear RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737
Catalog No. 50-114-9231