Learn More
MSC Dow Corning 99180484 10.1 oz Cartridge Black RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737
Catalog No. 501149233
Encompass
Dow Corning 99180484 10.1 oz Cartridge Black RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737
Catalog No. 50-114-9233 Supplier MSC Supplier No. 00231969
Dow Corning 99180484 10.1 oz Cartridge Black RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737